Learn More
Miele, Inc. E 403 Half Insert for 105 watch-glasses (Ø 50-60 mm)
Catalog No. 501713021
Encompass_Preferred
E 403 Mesh Basket
For optimum loading of up to 105 watch glasses.
Suitable for 105 watch glasses Ø 50 - 60 mm
Equipped with 36 bars at a distance of 0 3/8 (9) mm
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3021 Supplier Miele, Inc. Supplier No. 03830430
E 403 Mesh Basket
For optimum loading of up to 105 watch glasses.
Suitable for 105 watch glasses Ø 50 - 60 mm
Equipped with 36 bars at a distance of 0 3/8 (9) mm
Visit www.mieleusa.com/professional for technical information.