Learn More
Enzo Life Sciences E-4031 . dihydrochloride (5mg). CAS: 113558-89-7
SDP
Catalog No. 502011071
Encompass_Preferred
Potent and selective blocker of the inactivating inward rectifying potassium current IKR (HERG channel). Class III antiarrythmic agent with differential atrial vs. ventricular activity. Alternative name: 4'-[[1-[2-(6-methyl-2-pyridyl)ethyl]-4-piperidinyl]carbonyl]-methanesulfonanilide. Purity: ≥98% (HPLC). Solubility: Soluble in water (100mg/ml) or DMSO. Long Term Storage: Ambient.
Catalog No. 50-201-1071 Supplier Enzo Life Sciences Supplier No. BMLKC1580005
Potent and selective blocker of the inactivating inward rectifying potassium current IKR (HERG channel). Class III antiarrythmic agent with differential atrial vs. ventricular activity. Alternative name: 4'-[[1-[2-(6-methyl-2-pyridyl)ethyl]-4-piperidinyl]carbonyl]-methanesulfonanilide. Purity: ≥98% (HPLC). Solubility: Soluble in water (100mg/ml) or DMSO. Long Term Storage: Ambient.