Learn More
Cayman Chemical E-4031hydrochlorIde 10mg
SDP
Catalog No. 502972815
Encompass_Preferred
A methanesulfonanilide class III antiarrhythmic agent that prolongs cardiac action potential duration by blocking ERG K+ channels (IC50 = 29 nM); inhibits the ERG1, ERG2, and ERG3 isoforms with KD values of 99, 116, and 193 nM, respectively
Catalog No. 50-297-2815 Supplier Cayman Chemical Supplier No. 1520310 MG
A methanesulfonanilide class III antiarrhythmic agent that prolongs cardiac action potential duration by blocking ERG K+ channels (IC50 = 29 nM); inhibits the ERG1, ERG2, and ERG3 isoforms with KD values of 99, 116, and 193 nM, respectively