Learn More
Miele, Inc. E 408 Mobile unit TA drying co
Catalog No. 502870768
Encompass_Preferred
E 408 Mobile unit TA drying connection for 96 pipettes up to 450mm long
Catalog No. 50-287-0768 Supplier Miele, Inc. Supplier No. 3810130
Add to Cart
Add to Cart
E 408 Mobile unit TA drying connection for 96 pipettes up to 450mm long