Learn More
Cayman Chemical (E)-5-(2-Bromovinyl)uracil, Purity: ≥95%, CAS: 69304-49-0, 100 mg
SDP
Catalog No. 502983084
Encompass_Preferred
A pyrimidine base; is an inactive metabolite of the antiviral agents sorivudine and BVDU that may be regenerated to BVDU in vivo; irreversibly inactivates DPD in an NADPH-dependent manner; enhances the efficacy of the chemotherapeutic agent and DPD substrate 5-FU in a P388 murine leukemia model at 200 µmol/kg, increasing survival time
Catalog No. 50-298-3084 Supplier Cayman Chemical Supplier No. 21030100 MG
A pyrimidine base; is an inactive metabolite of the antiviral agents sorivudine and BVDU that may be regenerated to BVDU in vivo; irreversibly inactivates DPD in an NADPH-dependent manner; enhances the efficacy of the chemotherapeutic agent and DPD substrate 5-FU in a P388 murine leukemia model at 200 µmol/kg, increasing survival time