Learn More
Medchemexpress LLC (E)-5-(2-(quinolin-4-yl)vinyl)benzene-1,3-diol | 1016897-10-1 | 98.1% | 263.29 g/mol | C17H13NO2 | 100MG
SDP
Catalog No. 5000239654
Encompass_Preferred
RV01 is a small-molecule resveratrol analogue that inhibits acetaldehyde dehydrogenase (ALDH) and has been reported to reduce DNA damage; it is intended for biochemical and pharmacology research applications. The compound is supplied as a light-yellow powder (also available as DMSO solutions), has a molecular weight of 263.29 g/mol, and a typical purity of 98.1%; follow recommended storage conditions for stability.
Catalog No. 50-002-39654 Supplier Medchemexpress LLC Supplier No. HY126241100MG
RV01 is a small-molecule resveratrol analogue that inhibits acetaldehyde dehydrogenase (ALDH) and has been reported to reduce DNA damage; it is intended for biochemical and pharmacology research applications. The compound is supplied as a light-yellow powder (also available as DMSO solutions), has a molecular weight of 263.29 g/mol, and a typical purity of 98.1%; follow recommended storage conditions for stability.