Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-5-(4-propoxybenzylidene)thiazolidine-2,4-dione | 587852-28-6 | MFCD31382191 | 98.0% | 263.31 g·mol⁻¹ | C13H13NO3S | 100 MG
SDP

Catalog No. 5000248909
Encompass_Preferred

SMI-16a (CAS 587852-28-6) is a selective, ATP-competitive Pim kinase inhibitor used as a research tool to probe Pim kinase signaling and downstream cellular effects. It shows potent biochemical inhibition of Pim isoforms and measurable activity in cellular assays, and is supplied with analytical data for use in preclinical studies.

  • Selective Pim1 and Pim2 inhibitor.
  • Biochemical IC50: Pim1 0.15 μM, Pim2 0.02 μM; cellular activity in PC3 cells 48 μM.
  • Chemical formula C13H13NO3S and molecular weight 263.31 g·mol⁻¹.
  • Purity 98.0% as specified by the supplier.
  • High solubility in DMSO (≥ 100 mg/mL) and defined in vivo formulation options.
  • Powder storage stability: -20°C for 3 years, 4°C for 2 years.

Catalog No. 50-002-48909 Supplier Medchemexpress LLC Supplier No. HY101947100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SMI-16a (CAS 587852-28-6) is a selective, ATP-competitive Pim kinase inhibitor used as a research tool to probe Pim kinase signaling and downstream cellular effects. It shows potent biochemical inhibition of Pim isoforms and measurable activity in cellular assays, and is supplied with analytical data for use in preclinical studies.

  • Selective Pim1 and Pim2 inhibitor.
  • Biochemical IC50: Pim1 0.15 μM, Pim2 0.02 μM; cellular activity in PC3 cells 48 μM.
  • Chemical formula C13H13NO3S and molecular weight 263.31 g·mol⁻¹.
  • Purity 98.0% as specified by the supplier.
  • High solubility in DMSO (≥ 100 mg/mL) and defined in vivo formulation options.
  • Powder storage stability: -20°C for 3 years, 4°C for 2 years.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.