Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-5-((8-methoxy-2-methylquinolin-4-yl)amino)-N'-(4-(methylthio)benzylidene)-1H-indole-2-carbohydrazid | 2469975-55-9 | 98.6% | C28H25N5O2S | 10MG
SDP

Catalog No. 5000258680
Encompass_Preferred

Nur77 modulator 1 is a small-molecule research compound that modulates the nuclear receptor Nur77 (NR4A1), promotes Nur77-dependent endoplasmic reticulum stress and autophagy, and can induce apoptosis; it has been applied in hepatoma models and shows a measured binding affinity to Nur77.

  • Chemical formula C28H25N5O2S.
  • Molecular weight 495.60 g·mol⁻¹.
  • Purity 98.6%.
  • CAS number 2469975-55-9.
  • Reported KD against Nur77 3.58 μM.
  • Available in mg-scale quantities for research use.

Catalog No. 50-002-58680 Supplier Medchemexpress LLC Supplier No. HY13136410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nur77 modulator 1 is a small-molecule research compound that modulates the nuclear receptor Nur77 (NR4A1), promotes Nur77-dependent endoplasmic reticulum stress and autophagy, and can induce apoptosis; it has been applied in hepatoma models and shows a measured binding affinity to Nur77.

  • Chemical formula C28H25N5O2S.
  • Molecular weight 495.60 g·mol⁻¹.
  • Purity 98.6%.
  • CAS number 2469975-55-9.
  • Reported KD against Nur77 3.58 μM.
  • Available in mg-scale quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.