Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-5-fluoro-N-(3-(3-(hydroxyamino)-3-oxoprop-1-en-1-yl)-4-methoxybenzyl)-2-morpholinoben | 3031349-25-1 | 95.0% | 429.44 | 25 MG
SDP

Catalog No. 5000363477
Encompass_Preferred

This potent HDAC inhibitor demonstrates IC50s for HDAC1, HDAC2, HDAC3, HDAC6, and HDAC8, and inhibits cell migration and invasion. It induces apoptosis and cell cycle arrest at the G2/M phase, also showing anti-tumor activity.

  • Potent inhibition of HDAC1, HDAC2, HDAC3, HDAC6, and HDAC8.
  • Inhibits cell migration and invasion, and induces apoptosis and cell cycle arrest at G2/M phase.
  • Demonstrates anti-tumor activity.
  • Inhibits tubulin polymerization.
  • Increases expression of acetylated-α-tubulin and acetyl-histone H3.
  • Modulates mitochondrial membrane potential and reactive oxygen species levels.
  • Exhibits favorable liver microsomal stability.
  • Shows antiproliferative activities in various cancer cell lines.
  • Demonstrates antivascular activity and good cardiovascular safety in vivo.
  • Reduces tumor weights in allograft mouse models.

Catalog No. 50-003-63477 Supplier Medchemexpress LLC Supplier No. 25MGHY149029
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This potent HDAC inhibitor demonstrates IC50s for HDAC1, HDAC2, HDAC3, HDAC6, and HDAC8, and inhibits cell migration and invasion. It induces apoptosis and cell cycle arrest at the G2/M phase, also showing anti-tumor activity.

  • Potent inhibition of HDAC1, HDAC2, HDAC3, HDAC6, and HDAC8.
  • Inhibits cell migration and invasion, and induces apoptosis and cell cycle arrest at G2/M phase.
  • Demonstrates anti-tumor activity.
  • Inhibits tubulin polymerization.
  • Increases expression of acetylated-α-tubulin and acetyl-histone H3.
  • Modulates mitochondrial membrane potential and reactive oxygen species levels.
  • Exhibits favorable liver microsomal stability.
  • Shows antiproliferative activities in various cancer cell lines.
  • Demonstrates antivascular activity and good cardiovascular safety in vivo.
  • Reduces tumor weights in allograft mouse models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.