Learn More
TARGETMOL CHEMICALS INC E-6-DEHYDROPARADOL 10MG
SDP
Catalog No. 502581348
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (E)-[6]-Dehydroparadol ((6)-Dehydroparadol) an oxidative metabolite of [6]-Shogaol is a potent Nrf2 activator. purity: 99%
Catalog No. 50-258-1348 Supplier TARGETMOL CHEMICALS INC Supplier No. T1343610MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (E)-[6]-Dehydroparadol ((6)-Dehydroparadol) an oxidative metabolite of [6]-Shogaol is a potent Nrf2 activator. purity: 99%