Learn More
Apexbio Technology LLC E-64 66701-25-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064212
Encompass_Preferred
E-64 is a selective inhibitor targeting cysteine proteases isolated originally from Aspergillus cultures Structurally classified as an L-trans-epoxysuccinyl peptide it irreversibly inhibits proteases such as papain ficin and bromelain by covalent binding to the active-site cysteine residue Its inhibitory activity additionally extends to mammalian cysteine proteases including cathepsins B H and L as well as calcium-dependent protease calpain with reported IC50 values ranging approximately between 10-100 nM depending on specific enzyme assays and conditions In research laboratories E-64 is primarily utilized for mechanistic studies active-site titration assays and quantitative evaluation of cysteine protease concentrations and enzyme kinetics
Catalog No. 50-000-64212 Supplier Apexbio Technology LLC Supplier No. A25765.1
E-64 is a selective inhibitor targeting cysteine proteases isolated originally from Aspergillus cultures Structurally classified as an L-trans-epoxysuccinyl peptide it irreversibly inhibits proteases such as papain ficin and bromelain by covalent binding to the active-site cysteine residue Its inhibitory activity additionally extends to mammalian cysteine proteases including cathepsins B H and L as well as calcium-dependent protease calpain with reported IC50 values ranging approximately between 10-100 nM depending on specific enzyme assays and conditions In research laboratories E-64 is primarily utilized for mechanistic studies active-site titration assays and quantitative evaluation of cysteine protease concentrations and enzyme kinetics