Learn More
AG Scientific Inc E-64, 25 MG
Catalog No. 502236982
Encompass_Preferred
E-64 is an Irreversible, potent and highly selective inhibitor of cysteine proteases. It does not affect cysteine residues in other enzymes. E64 acts by forming a thioether bond with the thiol of the active cysteine. E-64 will not inhibit serine proteases (except trypsin) but inhibits activation-induced programmed cell death and restores defective immune responses in HIV. Specific active site titrant.CAS Number: 66701-25-5Molecular Weight: 357.4Chemical Formula: C15H27N5O5Solubility: Up to 20mg/ml in water, Up to 25mg/ml in DMSOStorage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6982 Supplier AG Scientific Inc Supplier No. E203025MG
E-64 is an Irreversible, potent and highly selective inhibitor of cysteine proteases. It does not affect cysteine residues in other enzymes. E64 acts by forming a thioether bond with the thiol of the active cysteine. E-64 will not inhibit serine proteases (except trypsin) but inhibits activation-induced programmed cell death and restores defective immune responses in HIV. Specific active site titrant.CAS Number: 66701-25-5Molecular Weight: 357.4Chemical Formula: C15H27N5O5Solubility: Up to 20mg/ml in water, Up to 25mg/ml in DMSOStorage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.