Learn More
Enzo Life Sciences E-64 (25mg). CAS: 66701-25-5
SDP
Supplier:  Enzo Life Sciences ALX260007M025
Encompass_Preferred
Protease inhibitor. Irreversible inhibitor of cysteine proteinases like papain, calpain, cathepsin B, L and H. Acts by forming a thioether bond with the thiol of the active cysteine. Does not inhibit serine proteinases. Inhibits degradation of autophagic cargo in autophagolysosomes. Alternative name: N-[N-(L-3-trans-Carboxyoxirane-2-carbonyl]-L-leucyl)-agmatine, L-trans-Epoxysuccinyl-leucylamide-(4-guanido)-butane. Formula: C15H27N5O5 . 0.5 H20. MW: 357.4. Purity: ≥98%. Appearance: White to off-white solid. Solubility: Soluble in water/ethanol 1:1 mixture (15 mg/ml). Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. 50-200-8219