Learn More
Research Products International Corp E-64 [Epoxysuccinyl-L-Leucylamido (4-guanido) butane], 5 Milligrams
SDP
Catalog No. 50488653
Encompass_Preferred
Irreversible cysteine protease inhibitor of calpain, cathepsin (B,H,L,S), papain and other cysteine proteases. Working concentration: 1-10uM.
Catalog No. 50-488-653 Supplier Research Products International Corp Supplier No. E570500.005
Add to Cart
Add to Cart
Irreversible cysteine protease inhibitor of calpain, cathepsin (B,H,L,S), papain and other cysteine proteases. Working concentration: 1-10uM.