Learn More
Medchemexpress LLC HY-15282 50mg , E-64 CAS:66701-25-5 Purity:>98%
SDP
Catalog No. 502259744
Encompass_Preferred
HY-15282 50mg E-64 CAS: 66701-25-5 E-64 (Proteinase inhibitor E 64) is a potent irreversible inhibitor against general cysteine proteases with IC50 of 9 nM for papain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9744 Supplier Medchemexpress LLC Supplier No. HY1528250MG
HY-15282 50mg E-64 CAS: 66701-25-5 E-64 (Proteinase inhibitor E 64) is a potent irreversible inhibitor against general cysteine proteases with IC50 of 9 nM for papain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.