Learn More
Medchemexpress LLC HY-15282 5mg Medchemexpress, E-64 CAS:66701-25-5 Purity:>98%
SDP
Catalog No. 502030604
Encompass_Preferred
Medchemexpress, HY-15282 5mg E-64 CAS:66701-25-5 E-64 (Proteinase inhibitor E 64) is a potent irreversible inhibitor against general cysteine proteases with IC50 of 9 nM for papain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0604 Supplier Medchemexpress LLC Supplier No. HY152825MG
Medchemexpress, HY-15282 5mg E-64 CAS:66701-25-5 E-64 (Proteinase inhibitor E 64) is a potent irreversible inhibitor against general cysteine proteases with IC50 of 9 nM for papain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.