Learn More
Selleck Chemical LLC E-64
SDP
Supplier:  Selleck Chemical LLC S7379250MG
Encompass_Preferred
E-64 is an irreversible and selective cysteine protease inhibitor and also inhibits papain calpain and cathepsins B and H but not serine proteases or aspartic proteases The IC50 for papain is 9 nM E-64 induces oxidative stress and apoptosis in Filarial Parasite
Catalog No. 50-000-04551