Learn More
Enzo Life Sciences E-64-C (5mg). CAS: 76684-89-4
SDP
Supplier:  Enzo Life Sciences BMLPI1060005
Encompass_Preferred
Cell-impermeable epoxysuccinyl peptide inhibitor of calpain and other cysteine proteases. Inhibits calpain and cathepsins B, H, and L. Alternative name: 2S,3S-trans-(Carboxyoxirane-2-carbonyl) -L-leucine-(3-methylbutyl) amide. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (5mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1442