Learn More
Crescent Chemical Co Inc E-64 RESEARCH GRADE
SDP
Catalog No. 50247574
Encompass_Preferred
E-64 research grade Inhibitor for papain and other cysteine (thiol) proteases. Effects on metastasis formation in mice (5). Size - 5MG Storage Conditions - -15 °C TO -25 °C Catalog Number - 21100.01 CAS 66701-25-5
Catalog No. 50-247-574 Supplier Crescent Chemical Co Inc Supplier No. SE21100.01
E-64 research grade Inhibitor for papain and other cysteine (thiol) proteases. Effects on metastasis formation in mice (5). Size - 5MG Storage Conditions - -15 °C TO -25 °C Catalog Number - 21100.01 CAS 66701-25-5