Learn More
Crescent Chemical Co Inc E-64 RESEARCH GRADE
Supplier: Crescent Chemical Co Inc SE21100.02
E-64 research grade Inhibitor for papain and other cysteine (thiol) proteases. Effects on metastasis formation in mice (5). Size - 25MG Storage Conditions - -15 °C TO -25 °C Catalog Number - 21100.02 CAS 66701-25-5