Learn More
AG Scientific Inc E-64d, 1 MG
Supplier:  AG Scientific Inc E24011MG
Encompass_Preferred
An irreversible, highly selective cysteine protease inhibitor and cell permeable analog of E-64D. Specifically E-64D is an irreversible cathepsin L and B inhibitor.E-64d has played an imporant role in studies of the Ebola virus (EBOV). E-64d has been shown to lead to the accumulation of CTFs and AICD with no change in gamma and beta secretase activities. Additionally, utilizing E-64d has shown results in accumulation of autophagic bodies inside vacuoles in Arabidopsis root cells.CAS Number: 88321-09-09Molecular Weight: 342.433Chemical Formula: C17H30N2O5Solubility: Soluble in EtOH (10 mg/mL) and DMSO (20 mg/mL)Storage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-7014