Learn More
Medchemexpress LLC E-7386 | 1799824-08-0 | 99.11% | 725.85 | 50 MG
SDP
Catalog No. 5000343804
Encompass_Preferred
E-7386 is an orally active CBP/beta-catenin modulator. It can induce T cell infiltration into tumors and enhance the antitumor activity of anti-PD-1 mAb in Wnt1 tumor syngeneic mice models. It demonstrates significant antitumor activity in the Wnt1 model, with clear T cell infiltration into tumors observed in the treatment group compared to the limited infiltration in the vehicle control group.
  • Orally active CBP/beta-catenin modulator.
  • Induces T cell infiltration into tumors.
  • Enhances antitumor activity of anti-PD-1 mAb in Wnt1 tumor syngeneic mice models.
  • Shows significant antitumor activity in Wnt1 models.
Catalog No. 50-003-43804 Supplier Medchemexpress LLC Supplier No. HY11138610MM/1ML
E-7386 is an orally active CBP/beta-catenin modulator. It can induce T cell infiltration into tumors and enhance the antitumor activity of anti-PD-1 mAb in Wnt1 tumor syngeneic mice models. It demonstrates significant antitumor activity in the Wnt1 model, with clear T cell infiltration into tumors observed in the treatment group compared to the limited infiltration in the vehicle control group.
  • Orally active CBP/beta-catenin modulator.
  • Induces T cell infiltration into tumors.
  • Enhances antitumor activity of anti-PD-1 mAb in Wnt1 tumor syngeneic mice models.
  • Shows significant antitumor activity in Wnt1 models.