Learn More
AdipoGen E-8-Methyl-6-nonenoic acid
SDP
Catalog No. 502560477
Encompass_Preferred
Chemical. CAS 59320-77-3. Formula C10H18O2. MW 170.25. Synthetic Intermediate of the capsaicin biosynthesis. Fatty acid derivative from leucine/valine pathway. Plays a crucial role in determining the efficacy of capsaicin levels. The quantity of E-8-Methyl-6-nonenoic acid determines the pungency in placental tissues of Capsicum.
Catalog No. 50-256-0477 Supplier AdipoGen Supplier No. CDXM0209M500
Chemical. CAS 59320-77-3. Formula C10H18O2. MW 170.25. Synthetic Intermediate of the capsaicin biosynthesis. Fatty acid derivative from leucine/valine pathway. Plays a crucial role in determining the efficacy of capsaicin levels. The quantity of E-8-Methyl-6-nonenoic acid determines the pungency in placental tissues of Capsicum.