Learn More
Cayman Chemical (E)-8-Methyl-6-nonenoic Acid, Purity: ≥95% (may contain up to 15% of the Z-isomer), CAS: 59320-77-3, 10 mg
SDP
Catalog No. 502965483
Encompass_Preferred
A potential thermal degradant of capsaicin with possible mutagenic effects
Catalog No. 50-296-5483 Supplier Cayman Chemical Supplier No. 1660910 MG
A potential thermal degradant of capsaicin with possible mutagenic effects