Learn More
Medchemexpress LLC (E)-9-amino-10-methyl-2-(naphthalen-1-ylmethylene)-1-oxo-1,2,3,4-tetrahydroacridin-10-ium iodide | 2252162-81-3 | 98.1% | 492.35 g/mol | C25H21IN2O | 5 MG
SDP
Catalog No. 5000221301
Encompass_Preferred
Aβ/tau aggregation-IN-1 is a small-molecule research compound that inhibits aggregation of amyloid-β (Aβ1-42) and tau proteins. It reduces β-sheet formation through noncovalent interactions, has reported brain permeability, and is supplied with documented solubility and storage recommendations for both in vitro and in vivo use.
  • Inhibits Aβ1-42 and tau aggregation in biochemical assays.
  • Reduces β-sheet formation via noncovalent interactions.
  • Reported to be brain permeable for central nervous system studies.
  • Suitable for in vitro and in vivo research applications.
  • High purity compound (reported >98%).
  • Soluble in DMSO; documented in vivo formulation options yield ≥1 mg/mL.
  • Store powder at 4°C; store solutions at -80°C (up to 6 months) or -20°C (up to 1 month).
Catalog No. 50-002-21301 Supplier Medchemexpress LLC Supplier No. HY1416615MG
Aβ/tau aggregation-IN-1 is a small-molecule research compound that inhibits aggregation of amyloid-β (Aβ1-42) and tau proteins. It reduces β-sheet formation through noncovalent interactions, has reported brain permeability, and is supplied with documented solubility and storage recommendations for both in vitro and in vivo use.
  • Inhibits Aβ1-42 and tau aggregation in biochemical assays.
  • Reduces β-sheet formation via noncovalent interactions.
  • Reported to be brain permeable for central nervous system studies.
  • Suitable for in vitro and in vivo research applications.
  • High purity compound (reported >98%).
  • Soluble in DMSO; documented in vivo formulation options yield ≥1 mg/mL.
  • Store powder at 4°C; store solutions at -80°C (up to 6 months) or -20°C (up to 1 month).