Learn More
Miele, Inc. E 943/2 Module for narrow neck
Catalog No. 502870775
Encompass_Preferred
E 943/2 Module for narrow necked glassware 100-500ml
Catalog No. 50-287-0775 Supplier Miele, Inc. Supplier No. 7459400
E 943/2 Module for narrow necked glassware 100-500ml