Learn More
Miele, Inc. E 960/1 Half Insert for wide-n
Catalog No. 502870737
Encompass_Preferred
E 960/1 Half Insert for wide-necked Erlenmeyrer flasks with 20 large+26 small spring hooks
Catalog No. 50-287-0737 Supplier Miele, Inc. Supplier No. 5892360
E 960/1 Half Insert for wide-necked Erlenmeyrer flasks with 20 large+26 small spring hooks