Learn More
Miele, Inc. E 965 Half Insert for 250-600m
Catalog No. 502870716
Encompass_Preferred
E 965 Half Insert for 250-600mL beakers with 15 x 3 holders
Catalog No. 50-287-0716 Supplier Miele, Inc. Supplier No. 5848290
E 965 Half Insert for 250-600mL beakers with 15 x 3 holders