Learn More
3M™ E-A-R™ E-Z-Fit™ Ear Plugs
Catalog No. 17987135A Shop All 3M Products
Change view
Click to view available options
Model:
312-1208
312-1222
Corded:
Corded
Uncorded
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Corded
17-987-135A 312-1222 Corded
17-987-135 312-1208 Uncorded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-987-135A Supplier 3M™ Supplier No. 7000127211
E-Z-Fit™ ear plugs are sized for smaller ears and bell shaped for easier insertion and removal on the job. Soft, slow-recovery foam comfortably expand in the ear canal to help provide reliable hearing protection.
  • Ideal for smaller ear canals
  • Poly bag keeps each pair of ear plugs clean before use
  • Test compatible with 3M™ E-A-Rfit™ Validation System
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
Cord Color Blue
Cord Material Vinyl
CSA Z94.2 Class AL
Size Small
For Use With (Application) Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
High Visibility Yes
Model 312-1222
Packaging Type Poly bag
Pairs per Package 200 Pairs/Pk.
Plug Material Foam
Plug Color Yellow
Noise Reduction Rating (NRR) 28 dB
Corded Corded
Plug Shape Bell
Disposable Single-use
Metal Detectable No