Learn More
3M™ E-A-R™ Flexible Fit High Attenuation Ear Plugs
Catalog No. 19072193 Shop All 3M Products
Change view
Click to view available options
Model:
328-1001
Corded:
Corded
Catalog No. Model Corded
19-072-193 328-1001 Corded
1 options
Catalog No. 19-072-193 Supplier 3M™ Supplier No. 7100212746
E-A-R™ Flexible Fit High Attenuation (HA) ear plugs have a soft foam tip and a flexible fitting stem that allows easy insertion using one or two hands. Suitable for activities that require gloves, cause dirty hands, or other situations when only one hand is free to fit ear plugs.
  • Soft foam stem for secure grip
  • No roll down helps keep ear tip clean
  • Ideal for workers with gloved or dirty hands or for situations when only one hand is free
  • Compatible with the 3M™ E-A-Rfit™ Dual-ear Validation System
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
Cord Color Blue
Cord Material Vinyl
CSA Z94.2 Class AL
Size Regular
For Use With (Application) Assembly, Blasting, Cleaning, Demolition, Facility Maintenance, Grinding, Machine Operations, Painting, Paint Preparation, Sanding, Welding
High Visibility Yes
Model 328-1001
Packaging Type Poly bag
Pairs per Package 100 Pairs/Pk.
Plug Material Elastomeric polymer
Plug Color Yellow
Noise Reduction Rating (NRR) 30 dB (two hand), 25 dB (one hand)
Corded Corded
Plug Shape Cone
Disposable Single-use
Metal Detectable No