Learn More
3M™ E-A-R™ One Touch™ Pro Ear Plug Dispenser
Catalog No. 19122409 Shop All 3M Products
Change view
Click to view available options
Disposable:
Reusable
Catalog No. Disposable
19-122-409 Reusable
1 options
Catalog No. 19-122-409 Supplier 3M™ Supplier No. 7100064963
  • $154.81
    $130.65 / Case of 1
    Save $24.16
    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
One Touch™ pro ear plug dispenser gives workers convenient access to hearing protection products. Ear plugs are dispensed one at a time with each turn to minimize spillage and waste.
  • Simple turn drops ear plug into a worker's hand one at a time
  • Drained catch tray, wind side shields, and durable plastic construction safeguards ear plugs in outdoor locations
  • Versatile design allows for countertop placement or wall mounting
  • Bulk style dispenser eliminates waste from individually wrapped ear plugs
  • Accepts a variety of different 3M™ Ear plugs in refill bottles
  • Refill bottle not included
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Ear Plug Accessibilty to Workers and Facility Visitors in Reception Areas and Noisy Environments
Holds 400 to 500 pairs of ear plugs
Model 391-0000
Product Type One Touch™ Pro Ear Plug Dispenser
Disposable Reusable
Material ABS plastic
Mounting Wall Mountable, Free-standing