Learn More
3M™ E-A-R™ UltraFit™ Silicone Ear Plugs
Catalog No. 19123999 Shop All 3M Products
Change view
Click to view available options
Corded ear plugs; Plug Material: Silicone; Cord Material: Cloth
Catalog No.
19-123-999 Corded ear plugs; Plug Material: Silicone; Cord Material: Cloth
1 options
This item is not returnable. View return policy
Catalog No. 19-123-999 Supplier 3M™ Supplier No. 7100239692
This item is not returnable. View return policy
UltraFit™ Silicone ear plugs offer convenient, corded comfort. Made from durable, medical-grade silicone this ear plug includes a cloth cord and helps provide in-ear comfort and convenience with a Noise Reduction Rating (NRR) of 25 dB.
  • Soft material easily adapts to the ear canal
  • Durable, medical-grade silicone is washable and reusable helping to reduce waste and costs
  • Three-flange design fits a wide range of ear sizes
  • No need to roll ear plugs before fitting so the tips stay clean even if hands are dirty
  • Noise reduction rating (NRR) 25 dB. CSA Class AL
  • Carrying case helps keep ear plugs clean and accessible for re-use
  • Cloth cord for comfort and convenience
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
Cord Color Red
Cord Material Cloth
CSA Z94.2 Class AL
Size Regular
For Use With (Application) Composite Finishing, Final Finish, Paint Preparation, Painting, Welding
High Visibility No
Model 340-3002
Pairs per Package 80 Pairs/Pk.
Plug Material Silicone
Noise Reduction Rating (NRR) 25 dB
Corded Corded
Plug Shape Flanged
Disposable Reusable
Metal Detectable No
Includes Pocket-size case