Learn More
3M™ E-A-Rsoft™ SuperFit™ Ear Plugs
Catalog No. 19051437 Shop All 3M Products
Change view
Click to view available options
Model:
311-1254
Corded:
Corded
Catalog No. Model Corded
19-051-437 311-1254 Corded
1 options
Catalog No. 19-051-437 Supplier 3M™ Supplier No. 7000002307
Add to Cart
Add to Cart
E-A-Rsoft™ SuperFit™ ear plugs are made of an advanced foam formulation that provides a smooth texture for in-ear comfort and softness. The exclusive orange ring design provides a visual to help confirm proper insertion.
  • Soft, pliable foam offers comfort over an extended time
  • Orange fitting ring provides a guideline for proper fitting
  • Test compatible with 3M™ E-A-Rfit™ Validation System.
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
Cord Color Orange
Cord Material Vinyl
CSA Z94.2 Class AL
Size Regular
For Use With (Application) Assembly, Blasting, Cleaning, Composite Finishing, Demolition, Electrical, Facility Maintenance, Final Finish, Grinding, Machine Operations, Paint Preparation, Painting, Sanding, Welding
High Visibility Yes
Model 311-1254
Packaging Type Poly bag
Pairs per Package 2000 Pairs/Cs
Plug Material Foam
Plug Color Yellow, orange
Noise Reduction Rating (NRR) 33 dB
Corded Corded
Plug Shape Bullet
Disposable Single-use
Metal Detectable No