Learn More
Medchemexpress LLC (E)-AG 556 | 133550-41-1 | >98.0% | 336.39 g/mol | C20H20N2O3 | 25 MG
SDP
Catalog No. 5000212265
Encompass_Preferred
(E)-AG 556 is a research-grade small molecule that selectively inhibits epidermal growth factor receptor (EGFR) activity and has been reported to block lipopolysaccharide (LPS)-induced tumor necrosis factor-α (TNF-α) production. It is supplied as a solid powder for use in biochemical and cellular assays and is intended for research use only.
  • Selective EGFR inhibition for targeted signaling studies.
  • Reported to reduce LPS-induced TNF-α production in cellular models.
  • Powder form suitable for preparation of stock solutions and dosing.
  • High purity appropriate for biochemical and cellular assays.
  • Recommended long-term storage at -20°C to preserve stability.
Catalog No. 50-002-12265 Supplier Medchemexpress LLC Supplier No. HY10104125MG
(E)-AG 556 is a research-grade small molecule that selectively inhibits epidermal growth factor receptor (EGFR) activity and has been reported to block lipopolysaccharide (LPS)-induced tumor necrosis factor-α (TNF-α) production. It is supplied as a solid powder for use in biochemical and cellular assays and is intended for research use only.
  • Selective EGFR inhibition for targeted signaling studies.
  • Reported to reduce LPS-induced TNF-α production in cellular models.
  • Powder form suitable for preparation of stock solutions and dosing.
  • High purity appropriate for biochemical and cellular assays.
  • Recommended long-term storage at -20°C to preserve stability.