Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-ag 99 ((E)-tyrphostin 46) | 122520-85-8 | 98.4% | 204.19 | C10H8N2O3 | 1 ML
SDP

Catalog No. 5000211530
Encompass_Preferred

(E)-AG 99 ((E)-Tyrphostin 46) is a small-molecule tyrosine kinase inhibitor that potently inhibits the epidermal growth factor receptor (EGFR). It is supplied for research use and is available as dry powder or as a pre-dissolved 10 mM solution in DMSO for cell-based and biochemical assays.

  • Potent EGFR inhibitor suitable for kinase and cellular assays.
  • High purity (~98.4%) for reproducible experimental results.
  • Available as 10 mM solution in DMSO for immediate use.
  • Also available in multiple mass formats (5 mg-100 mg) for flexibility.
  • Molecular formula C10H8N2O3 and molecular weight 204.19 for dosing calculations.
  • CAS number 122520-85-8 provided for unambiguous substance identification.

Catalog No. 50-002-11530 Supplier Medchemexpress LLC Supplier No. HY10096210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E)-AG 99 ((E)-Tyrphostin 46) is a small-molecule tyrosine kinase inhibitor that potently inhibits the epidermal growth factor receptor (EGFR). It is supplied for research use and is available as dry powder or as a pre-dissolved 10 mM solution in DMSO for cell-based and biochemical assays.

  • Potent EGFR inhibitor suitable for kinase and cellular assays.
  • High purity (~98.4%) for reproducible experimental results.
  • Available as 10 mM solution in DMSO for immediate use.
  • Also available in multiple mass formats (5 mg-100 mg) for flexibility.
  • Molecular formula C10H8N2O3 and molecular weight 204.19 for dosing calculations.
  • CAS number 122520-85-8 provided for unambiguous substance identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.