Learn More
Medchemexpress LLC (E)-Akt inhibitor-IV | 959841-49-7 | 98.6% | 614.54 | 1 ML
SDP
Catalog No. 5000361840
Encompass_Preferred
(E)-Akt inhibitor-IV ((E)-AKTIV) is a PI3K-Akt inhibitor with potent cytotoxic properties. It is intended for research use only and is not sold to patients.
  • Acts as a PI3K-Akt inhibitor
  • Exhibits potent cytotoxic properties
  • Shows an average GI20 of 0.04 μM on four cell lines including MDA-MB468, MDA-MB231, MCF7, and 184B5 cells
  • Has little effect on the growth of 184B5 non-cancer cells at average GI20 doses
Catalog No. 50-003-61840 Supplier Medchemexpress LLC Supplier No. HY1497110MM/1ML
(E)-Akt inhibitor-IV ((E)-AKTIV) is a PI3K-Akt inhibitor with potent cytotoxic properties. It is intended for research use only and is not sold to patients.
  • Acts as a PI3K-Akt inhibitor
  • Exhibits potent cytotoxic properties
  • Shows an average GI20 of 0.04 μM on four cell lines including MDA-MB468, MDA-MB231, MCF7, and 184B5 cells
  • Has little effect on the growth of 184B5 non-cancer cells at average GI20 doses