Learn More
Medchemexpress LLC (E)-Aztreonam | 99341-02-3 | 98.68% | 435.43 | 5 MG
SDP
Catalog No. 5000364648
Encompass_Preferred
(E)-Aztreonam is a monocyclic beta-lactam antibiotic with potential for research into infections caused by gram-negative pathogens.
  • It is a monocyclic beta-lactam antibiotic.
  • It has potential for research of infections caused by gram-negative pathogens.
  • It can induce an increased level of cellular activation in peritoneal cells.
  • It can increase the capacity of IL-1 production by peritoneal macrophages in mice.
Catalog No. 50-003-64648 Supplier Medchemexpress LLC Supplier No. 5MGHY136502
(E)-Aztreonam is a monocyclic beta-lactam antibiotic with potential for research into infections caused by gram-negative pathogens.
  • It is a monocyclic beta-lactam antibiotic.
  • It has potential for research of infections caused by gram-negative pathogens.
  • It can induce an increased level of cellular activation in peritoneal cells.
  • It can increase the capacity of IL-1 production by peritoneal macrophages in mice.