Learn More
Carbosynth LLC. H-Glu[Cyclo(Arg-Ala-Asp-D-Phe-Lys)]2, 25mg, MW: 1346.52, Molecular formula: C61H91N19O16
SDP
Supplier:  Carbosynth LLC. PCI3979PI25MG
Encompass_Preferred
Synonyms: E-[c(RADfK)2], H-Glu[cyclo(Arg-Ala-Asp-D-Phe-Lys)]2, E-[c(RADfK)2], Application: Negative Control Peptide for PCI-3651-PI, Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20°C
Catalog No. 50-168-6364