Learn More
Bachem H-Glu[cyclo(-Arg-Gly-Asp-D-Phe-Lys)]-cyclo(-Arg-Gly-Asp-D-Phe-Lys)
SDP
Supplier:  Bachem 4086610.0005
Encompass_Preferred
H-Glu[cyclo(-Arg-Gly-Asp-D-Phe-Lys)]-cyclo(-Arg-Gly-Asp-D-Phe-Lys) Trifluoroacetate, 250612-47-6, C59H87N19O16, Mr 1318.46, 5mg. E(c(RGDfK))2 (SU 016), a dimeric form of c(RGDfK), showed improved tumor targeting properties compared to the monomer. E(c(RGDfK))2 can be easily conjugated to dyes, drug molecules, or chelators. The 68Ga complex of DOTA-E(c(RGDfK))2 has been evaluated as theranostic radiopharmaceutical.
Catalog No. 50-194-6559