Learn More
Abcam E-Cadherin Antibody, 100 μL, Recombinant Rabbit Monoclonal, Clone EP700Y, Liquid
Supplier: Abcam AB40772100UL
This E-Cadherin antibody is a recombinant rabbit monoclonal antibody (clone EP700Y) specifically engineered for the detection of human E-Cadherin, essential for intercellular junctions. It is designed for reliable consistency, enhanced sensitivity, and specificity across multiple immunoassay applications. The antibody has been validated for various techniques, making it a critical tool for researchers studying cell adhesion, epithelial differentiation, and related pathologies.
- Host species is rabbit
- Clonality is monoclonal
- Reactivity against human E-Cadherin
- Validated for Western Blotting (WB), IHC-Paraffin (IHC-P), ICC/IF, Flow Cytometry, and multiplex IHC (mIHC)
- Form is liquid
- Storage at -20°C to avoid freeze/thaw cycles
- Shipped at 4°C
- Observed band sizes in WB are 80 kDa, 97 kDa, 100 kDa, 120 kDa
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.