Learn More
Advanced Biomatrix E-Cadherin Human 0.1 MG
Catalog No. 502255902
Encompass_Preferred
E-Cadherin protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer.
Catalog No. 50-225-5902 Supplier Advanced Biomatrix Supplier No. 50850.1MG
E-Cadherin protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer.