Learn More
ECM Bioscience E-Cadherin (Cytoplasmic) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience CM1681
Encompass_Preferred
Clone (M168) was generated from a mouse recombinant E-Cadherin protein containing amino acids in the C-terminal region. This sequence is highly conserved in human and rat E-cadherin. Accession #P09803. This E-cadherin antibody detects a 120 kDa* protein in human A431 cells, and does not cross-react with VE-cadherin or N-cadherin. Applications: WB, E, IP, ICC, IHC; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Uvomorulin, Cadherin-1, CTF1, CTF2, CTF3, CD324, Epithelial Cadherin
Catalog No. 50-172-0717