Learn More
Medchemexpress LLC HY-N1378 5mg Medchemexpress, (E)-Cardamonin CAS:19309-14-9 Purity:>98%
SDP
Catalog No. 501933815
Encompass_Preferred
Medchemexpress, HY-N1378 5mg (E)-Cardamonin CAS:19309-14-9 (E)-Cardamonin ((E)-Cardamomin) is a novel antagonist of hTRPA1 cation channel with an IC 50 of 454 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3815 Supplier Medchemexpress LLC Supplier No. HYN13785MG
Medchemexpress, HY-N1378 5mg (E)-Cardamonin CAS:19309-14-9 (E)-Cardamonin ((E)-Cardamomin) is a novel antagonist of hTRPA1 cation channel with an IC 50 of 454 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.