Learn More
LGC GENOMICS LLC (Lucigen) E. cloni 10G Chemically Competent Cells, 4 platesClone, construct libraries, transform Gibson Assembly reactions all with high-yield, T1 phage-resistant cells in high efficiencies
Catalog No. 501047941
Encompass
Lucigens E. cloni competent cells share the most useful genetic elements of standard cloning strains like DH5™, DH10B™, JM109, TOP10, etc. and directly replace them in cloning protocols. E. cloni chemically competent cells provide the performance researchers need with ease of useonly common laboratory equipment is required. These cells provide solutions for a wide range of applications at economical prices. E. cloni 10G Competent Cells. Routine cloning, subcloning, and plasmid isolation with or without blue/white screening.
This item is not returnable. View return policy
Catalog No. 50-104-7941 Supplier LGC GENOMICS LLC Supplier No. 600964
Lucigens E. cloni competent cells share the most useful genetic elements of standard cloning strains like DH5™, DH10B™, JM109, TOP10, etc. and directly replace them in cloning protocols. E. cloni chemically competent cells provide the performance researchers need with ease of useonly common laboratory equipment is required. These cells provide solutions for a wide range of applications at economical prices. E. cloni 10G Competent Cells. Routine cloning, subcloning, and plasmid isolation with or without blue/white screening.