Learn More
LGC GENOMICS LLC (Lucigen) E. cloni 10G ELITE Electrocompetent Cells, 12 Rxns (SOLOs)Clone, construct libraries, transform Gibson Assembly reactions all with high-yield, T1 phage-resistant cells in high efficiencies
Catalog No. NC0749147
Encompass
Lucigens E. cloni competent cells share the most useful genetic elements of standard cloning strains like DH5™, DH10B™, JM109, TOP10, etc. and directly replace them in cloning protocols. However, E. cloni electrocompetent cells incorporate a unique manufacturing technology that increases transformation efficiency, recombinant yields and reliability, while decreasing costs. These cells provide solutions for a wide range of applications at economical prices. E. cloni 10G & 10GF ELITE Electrocompetent Cells > 2 10^10 cfu/g pUC DNA. ELITE Cells have twice the transformation efficiency compared to ultra high efficiency cells from other suppliers. ELITE Cells provide large numbers of transformants from hard-to-clone fragments or limited DNA at a lower price.
This item is not returnable. View return policy
Catalog No. NC0749147 Supplier LGC GENOMICS LLC Supplier No. 600511
Lucigens E. cloni competent cells share the most useful genetic elements of standard cloning strains like DH5™, DH10B™, JM109, TOP10, etc. and directly replace them in cloning protocols. However, E. cloni electrocompetent cells incorporate a unique manufacturing technology that increases transformation efficiency, recombinant yields and reliability, while decreasing costs. These cells provide solutions for a wide range of applications at economical prices. E. cloni 10G & 10GF ELITE Electrocompetent Cells > 2 10^10 cfu/g pUC DNA. ELITE Cells have twice the transformation efficiency compared to ultra high efficiency cells from other suppliers. ELITE Cells provide large numbers of transformants from hard-to-clone fragments or limited DNA at a lower price.