Learn More
LGC GENOMICS LLC (Lucigen) E. cloni 5-alpha Chemically Competent Cells, 24 Rxns (DUOs)Clone and subclone with high efficiency, chemically competent DH5-alpha cells.
Catalog No. NC2162033
Encompass
Direct replacements for standard cloning strainsOptimized genetics for high yields: endonuclease and recombination minus, for blue/white screening-capable.Competent Cells include Control DNA and Recovery Medium, and are packaged as DUOs (2 transformations per tube). Recovery Medium is also available separately. The specified transformation efficiencies are with pUC DNA, unless indicated otherwise.
This item is not returnable. View return policy
Catalog No. NC2162033 Supplier LGC GENOMICS LLC Supplier No. 606022
Direct replacements for standard cloning strainsOptimized genetics for high yields: endonuclease and recombination minus, for blue/white screening-capable.Competent Cells include Control DNA and Recovery Medium, and are packaged as DUOs (2 transformations per tube). Recovery Medium is also available separately. The specified transformation efficiencies are with pUC DNA, unless indicated otherwise.