Learn More
LGC GENOMICS LLC (Lucigen) E. cloni 10G CLASSIC Electrocomp Cells, 24 Rxns (SixPacks)Clone, construct libraries, transform Gibson Assembly reactions all with high-yield, T1 phage-resistant cells in high efficiencies
Supplier:  LGC GENOMICS LLC 601171
Encompass
Lucigens E. cloni competent cells share the most useful genetic elements of standard cloning strains like DH5™, DH10B™, JM109, TOP10, etc. and directly replace them in cloning protocols. However, E. cloni electrocompetent cells incorporate a unique manufacturing technology that increases transformation efficiency, recombinant yields and reliability, while decreasing costs. These cells provide solutions for a wide range of applications at economical prices. E. cloni 10G CLASSIC Electrocompetent Cells > 5 10^9 cfu/g pUC DNA. CLASSIC Cells are high efficiency cells with a substantially lower cost per reaction. These cells are the most economical choice for standard cloning and library construction. 10G CLASSIC Cells are available in larger package sizes for convenient use in higher volume cloning applications.
Catalog No. NC1725435