Learn More
LGC GENOMICS LLC (Lucigen) E. cloni EXPRESS BL21(DE3)Chemically Competent Cells,12 Rxns (DUOs)
Supplier:  LGC GENOMICS LLC 604011
Encompass
E. cloni EXPRESS BL21(DE3)Chemically Competent Cells,12 Rxns (DUOs)Get better colony counts for routine protein expression with any E. coli T7 expression vector, with this high-efficiency BL21(DE3) strain.Best value for routine protein expressionAvailable as Electrocompetent and Chemically Competent Cells.Each E. cloni EXPRESS Kit contains: the indicated E. cloni EXPRESS Electrocompetent or Chemically Competent Cells in DUO packaging (2 transformations per tube), Expression Recovery Medium (lactose minus), pUC19 Positive Control Plasmid, and complete protocols.
Catalog No. NC2160377