Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-CLX-0921 | 606932-81-4 | 98.2% | 519.57 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY19394A1MG

Encompass_Preferred

(E)-CLX-0921 is an orally active PPARγ agonist, an E-isomer of CLX-0921, with an IC50 of 1.54 μM. It demonstrates potent antihyperglycemic activity and is utilized in the study of type 2 diabetes.

  • Orally active PPARγ agonist
  • Potent antihyperglycemic activity
  • Used for the study of type 2 diabetes
  • Purity: 98.17%
  • Molecular weight: 519.57
  • Appearance: Solid
  • Color: White to off-white
  • Stable under recommended storage conditions
  • Shipped at room temperature (in continental US)
  • Stored at 4°C, sealed, away from moisture (solid)
  • Stored at -80°C for 6 months or -20°C for 1 month (in solvent)

Catalog No. 50-003-84010


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.