Learn More
Bioss E. coli
SDP
Supplier:  Bioss BSM72122M
Encompass_Preferred
E coli Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-64737